Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003145.3 | NP_003136.1 | 6 | 720 | ||
XM_011509923.2 | XP_011508225.1 | 6 | 745 | |||
GenBank mRNA | AK092473.1 | - | 2 | 1430 | ||
AK222600.1 | - | 6 | 663 | |||
AK300678.1 | - | 6 | 720 | |||
AY251536.1 | - | 6 | 680 | |||
BC000341.2 | - | 6 | 666 | 2822983 | ||
D37991.1 | - | 6 | 664 | |||
X74104.1 | - | 6 | 690 |