Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080675.3 | NP_542406.2 | 10 | 721 | ||
XM_011528573.1 | XP_011526875.1 | 11 | 805 | |||
XM_011528574.1 | XP_011526876.1 | 9 | 661 | |||
XM_011528575.1 | XP_011526877.1 | 7 | 440 | |||
GenBank mRNA | AF401350.1 | - | 10 | 721 | ||
BC026118.2 | - | 10 | 721 | 4824412 | ||
BC029528.1 | - | 10 | 714 | 5271065 |