Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PKD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138295.3 | NP_612152.1 | 8, 9 | 1273 | ||
XM_017011798.1 | XP_016867287.1 | 9, 10 | 7678 | |||
GenBank mRNA | AB061683.1 | - | 8, 9 | 1239 |