Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322880.1 | NP_001309809.1 | 3, 4 | 1004 | ||
NM_001322881.1 | NP_001309810.1 | 3, 4 | 910 | |||
NM_139167.3 | NP_631906.2 | 3, 4 | 1004 | |||
GenBank mRNA | AY028700.1 | - | 2, 3 | 305 | ||
AY502063.1 | - | 3, 4 | 344 | |||
BC069459.1 | - | 2, 3 | 305 |