Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144658.3 | NP_653259.3 | 7 | 726 | ||
XM_005262368.3 | XP_005262425.1 | 7 | 971 | |||
XM_005262369.4 | XP_005262426.1 | 7 | 665 | |||
XM_011531276.2 | XP_011529578.1 | 7 | 606 | |||
GenBank mRNA | AK124735.1 | - | 1 | 111 | ||
AY692226.1 | - | 7 | 726 |