Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145004.6 | NP_659441.3 | 11 | 1251 | ||
XM_006716298.2 | XP_006716361.1 | 11 | 1045 | |||
XM_011544433.1 | XP_011542735.1 | 11 | 1045 | |||
XM_017013183.1 | XP_016868672.1 | 10 | 976 | |||
XM_017013184.1 | XP_016868673.1 | 11 | 1045 | |||
XM_017013185.1 | XP_016868674.1 | 10 | 1051 | |||
XM_017013186.1 | XP_016868675.1 | 7 | 655 | |||
XM_017013187.1 | XP_016868676.1 | 3 | 191 | |||
XM_017013188.1 | XP_016868677.1 | 2 | 160 | |||
GenBank mRNA | AK131038.1 | - | 3 | 220 | ||
AK313864.1 | - | 11 | 1072 | |||
AY358739.1 | - | 11 | 1131 | |||
BC026085.2 | - | 11 | 1035 | 4838404 | ||
BC026169.2 | - | 11 | 1062 | 4829390 | ||
BC028702.2 | - | 11 | 1045 | 4829385 | ||
BC030014.2 | - | 11 | 1038 | 4830659 | ||
BC030698.2 | - | 11 | 1053 | 4822988 | ||
BC034975.2 | - | 11 | 1046 | 4829806 |