Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313994.1 | NP_001300923.1 | 20, 21 | 2162 | ||
NM_145004.6 | NP_659441.3 | 23, 24 | 2560 | |||
XM_006716298.2 | XP_006716361.1 | 22, 23 | 2315 | |||
XM_006716299.1 | XP_006716362.1 | 21, 22 | 2036 | |||
XM_011544434.1 | XP_011542736.1 | 21, 22 | 2135 | |||
XM_017013183.1 | XP_016868672.1 | 22, 23 | 2285 | |||
XM_017013184.1 | XP_016868673.1 | 20, 21 | 2237 | |||
XM_017013185.1 | XP_016868674.1 | 22, 23 | 2360 | |||
XM_017013186.1 | XP_016868675.1 | 19, 20 | 1964 | |||
XM_017013187.1 | XP_016868676.1 | 15, 16 | 1500 | |||
XM_017013188.1 | XP_016868677.1 | 14, 15 | 1469 | |||
GenBank mRNA | AK122608.1 | - | 21, 22 | 1915 | ||
AK131038.1 | - | 15, 16 | 1529 | |||
AK300148.1 | - | 20, 21 | 2120 | |||
AK313864.1 | - | 23, 24 | 2381 | |||
AY358739.1 | - | 23, 24 | 2440 | |||
BC026085.2 | - | 23, 24 | 2344 | 4838404 | ||
BC026169.2 | - | 23, 24 | 2371 | 4829390 | ||
BC028702.2 | - | 23, 24 | 2354 | 4829385 | ||
BC030014.2 | - | 23, 24 | 2347 | 4830659 | ||
BC030698.2 | - | 23, 24 | 2362 | 4822988 | ||
BC034975.2 | - | 23, 24 | 2355 | 4829806 |