Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145115.2 | NP_660090.2 | 8 | 2281 | ||
XM_005250194.2 | XP_005250251.1 | 6 | 2053 | |||
XM_011515905.2 | XP_011514207.1 | 8 | 2592 | |||
XM_011515906.2 | XP_011514208.1 | 9 | 2699 | |||
XM_011515907.2 | XP_011514209.1 | 7 | 2536 | |||
XM_011515908.2 | XP_011514210.1 | 8 | 2606 | |||
XM_017011824.1 | XP_016867313.1 | 8 | 2328 | |||
XM_017011825.1 | XP_016867314.1 | 8 | 2633 | |||
XM_017011826.1 | XP_016867315.1 | 9 | 2310 | |||
GenBank mRNA | AK057030.1 | - | 8 | 2258 | ||
AK160369.1 | - | 3 | 2853 | |||
BC089402.1 | - | 5 | 1823 | |||
BX648158.1 | - | 7 | 3664 |