Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDARADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080738.3 | NP_542776.1 | 6 | 2688 | ||
NM_145861.2 | NP_665860.2 | 6 | 2569 | |||
GenBank mRNA | AK096339.1 | - | 6 | 2688 | ||
BC046928.1 | - | 1 | 1510 |