Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282506.1 | NP_001269435.1 | 2 | 1755 | ||
NM_001291833.1 | NP_001278762.1 | 3 | 1230 | |||
NM_020790.1 | NP_065841.1 | 3 | 1290 | |||
NM_152673.3 | NP_689886.3 | 3 | 1242 | |||
GenBank mRNA | AB037780.1 | - | 2 | 3051 | ||
AB098731.1 | - | 2 | 1493 | |||
AK027314.1 | - | 4 | 993 | |||
AK075138.1 | - | 3 | 951 | |||
AK298316.1 | - | 3 | 1415 | |||
AK311504.1 | - | 3 | 1240 | |||
AY358449.1 | - | 3 | 1175 | |||
BC029267.1 | - | 3 | 1201 | 3851952 | ||
BC044243.1 | - | 3 | 1129 | 5754328 | ||
BM697386.1 | - | 1 | 342 |