Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152754.2 | NP_689967.2 | 1, 2 | 186 | ||
XM_011515960.1 | XP_011514262.1 | 3, 4 | 496 | |||
XM_017011873.1 | XP_016867362.1 | 2, 3 | 685 | |||
GenBank mRNA | AK309116.1 | - | 3, 4 | 496 | ||
AY358937.1 | - | 2, 3 | 252 |