Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126132.2 | NP_001119604.1 | 10 | 648 | ||
NM_001126133.2 | NP_001119605.1 | 9 | 615 | |||
NM_001291774.1 | NP_001278703.1 | 10 | 710 | |||
NM_003283.5 | NP_003274.3 | 10 | 648 | |||
XM_011527246.2 | XP_011525548.1 | 9 | 525 | |||
XM_017027186.1 | XP_016882675.1 | 11 | 721 | |||
XM_017027187.1 | XP_016882676.1 | 10 | 709 | |||
GenBank mRNA | AK291352.1 | - | 10 | 500 | ||
AK308199.1 | - | 9 | 467 | |||
AY762903.1 | - | 9 | 443 | |||
AY762904.1 | - | 8 | 410 | |||
BC010963.1 | - | 10 | 501 | 4291828 | ||
BC022086.2 | - | 10 | 549 | |||
BC034143.1 | - | 10 | 500 | 4328089 | ||
BC107798.1 | - | 9 | 470 | 4247379 | ||
BT019630.1 | - | 8 | 410 | |||
M19308.1 | - | 9 | 468 | |||
M19309.1 | - | 10 | 500 | |||
S51938.1 | - | 8 | 410 | |||
S69208.1 | - | 10 | 484 | |||
S69209.1 | - | 9 | 451 |