Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF520800.1 | - | 8 | 765 | ||
AF520805.1 | - | 6 | 640 | |||
AF548115.1 | - | 3 | 336 | |||
AF548116.1 | - | 4 | 422 | |||
AF548121.1 | - | 332 | ||||
BC113541.1 | - | 7 | 704 | 8322593 | ||
BC113543.1 | - | 7 | 704 | 8322595 | ||
BC143837.1 | - | 8 | 716 | 9052352 |
Gene Symbol: LOC107983965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011612.1 | XP_016867101.1 | 9 | 1062 |