Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244889.1 | NP_001231818.1 | 9 | 1862 | ||
NM_170744.4 | NP_734465.2 | 10 | 1895 | |||
XM_011539453.1 | XP_011537755.1 | 11 | 1444 | |||
XM_017015834.1 | XP_016871323.1 | 11 | 1454 | |||
XM_017015835.1 | XP_016871324.1 | 10 | 2247 | |||
GenBank mRNA | AB096256.1 | - | 10 | 1850 | ||
AK128132.1 | - | 10 | 1895 | |||
AY126437.1 | - | 9 | 1470 | |||
AY358351.1 | - | 10 | 1841 |