Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308360.1 | NP_001295289.1 | 11 | 2237 | ||
NM_170776.4 | NP_740746.4 | 12 | 2376 | |||
XM_006721170.3 | XP_006721233.1 | 9 | 1770 | |||
XM_011522954.1 | XP_011521256.1 | 8 | 1657 | |||
GenBank mRNA | AK172801.1 | - | 12 | 2376 | ||
BC064508.1 | - | 12 | 2362 | 6424664 |
Set Membership: |
![]() |