Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173560.3 | NP_775831.2 | 2 | 403 | ||
XM_011535589.1 | XP_011533891.1 | 2 | 398 | |||
GenBank mRNA | BC039248.1 | - | 2 | 403 | 5272028 | |
CA842450.1 | - | 2 | 334 |