Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173562.4 | NP_775833.2 | 3 | 669 | ||
XM_005248940.4 | XP_005248997.1 | 4 | 524 | |||
XM_006715023.2 | XP_006715086.1 | 3 | 442 | |||
XM_011514398.2 | XP_011512700.1 | 4 | 520 | |||
GenBank mRNA | AK304377.1 | - | 3 | 390 | ||
AL832381.1 | - | 2 | 181 | |||
AY305862.1 | - | 3 | 669 |