Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK32C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318878.1 | NP_001305807.1 | 6 | 879 | ||
NM_001318879.1 | NP_001305808.1 | 6 | 740 | |||
NM_173575.3 | NP_775846.2 | 6 | 841 | |||
NR_134911.1 | 6 | 518 | ||||
XM_011539688.1 | XP_011537990.1 | 6 | 912 | |||
XM_011539690.2 | XP_011537992.1 | 6 | 827 | |||
XM_011539693.2 | XP_011537995.1 | 6 | 845 | |||
XM_011539694.1 | XP_011537996.1 | 6 | 579 | |||
XM_011539695.1 | XP_011537997.1 | 6 | 780 | |||
XM_011539696.1 | XP_011537998.1 | 6 | 587 | |||
GenBank mRNA | AK057849.1 | - | 6 | 518 | ||
AK098366.1 | - | 5 | 678 | |||
AK302102.1 | - | 6 | 826 | |||
AK308956.1 | - | 6 | 918 | |||
AK310725.1 | - | 6 | 845 | |||
AY098866.1 | - | 6 | 841 | |||
BC045760.1 | - | 6 | 730 | 4831007 |