Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TXNRD1 SIRNA Assays ›
Gene Symbol:
TXNRD1
Gene Name:
thioredoxin reductase 1
Gene Aliases:
GRIM-12, TR, TR1, TRXR1, TXNR
Chromosome Location:
Chr.12: 104215779 - 104350307 on Build GRCh38
UniGene ID:
Hs.654922
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
12981
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TXNRD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001093771.2
NP_001087240.1
6
593
NM_001261445.1
NP_001248374.1
4
1015
NM_001261446.1
NP_001248375.1
3
570
NM_003330.3
NP_003321.3
4
930
NM_182729.2
NP_877393.1
4
693
NM_182742.2
NP_877419.1
3
820
NM_182743.2
NP_877420.1
3
583
GenBank mRNA
AF077367.1
-
3
247
AF208018.1
-
2
143
AJ001050.1
-
2
156
AK226071.1
-
4
401
AK293322.1
-
3
230
AK295067.1
-
4
329
AK296495.1
-
3
220
AK304241.1
-
6
576
AK308881.1
-
4
624
AK312262.1
-
4
592
AL701549.1
-
5
538
AY057105.1
-
6
569
AY344080.1
-
4
314
AY344081.1
-
4
363
AY344083.1
-
4
480
AY344084.1
-
3
210
AY344086.1
-
4
319
AY344087.1
-
4
384
AY344088.1
-
4
334
AY344089.1
-
4
335
AY344092.1
-
4
322
AY344093.1
-
4
306
AY344096.1
-
3
260
AY344669.1
-
3
191
AY344670.1
-
3
315
AY344672.1
-
3
281
AY344673.1
-
3
209
AY344675.1
-
3
178
AY344679.1
-
3
243
BC018122.1
-
4
306
3883490
BT019640.1
-
2
119
CR536506.1
-
2
119
D88687.1
-
4
371
DA319870.1
-
3
370
DA440625.1
-
4
395
S79851.1
-
4
558
X91247.1
-
4
558
Back To Top
More Information
Panther Classification:
Molecular Function -
reductase
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
mesoderm formation
selenium compound metabolic process
signal transduction
cell proliferation
nucleobase-containing small molecule interconversion
cell redox homeostasis
oxidation-reduction process
cellular oxidant detoxification
thioredoxin-disulfide reductase activity
protein binding
electron carrier activity
protein disulfide oxidoreductase activity
flavin adenine dinucleotide binding
methylselenol reductase activity
methylseleninic acid reductase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0