Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173846.4 | NP_776245.2 | 4 | 656 | ||
XM_005267324.4 | XP_005267381.1 | 4 | 717 | |||
XM_017020966.1 | XP_016876455.1 | 4 | 620 | |||
GenBank mRNA | AK098680.1 | - | 4 | 865 | ||
BC027610.1 | - | 3 | 918 | |||
BC038970.2 | - | 3 | 545 | 5169309 |