Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257378.1 | NP_001244307.1 | 14 | 1688 | ||
NM_001257379.1 | NP_001244308.1 | 13 | 1518 | |||
NM_001257382.1 | NP_001244311.1 | 13 | 1588 | |||
NM_174975.4 | NP_777635.1 | 12 | 1470 | |||
GenBank mRNA | AK298079.1 | - | 13 | 1351 | ||
AY240872.1 | - | 12 | 1470 | |||
BC101001.2 | - | 14 | 1627 | 40006372 | ||
BC101002.1 | - | 13 | 1457 | 40006375 | ||
BC101003.1 | - | 13 | 1527 | 40006376 | ||
BC101004.1 | - | 12 | 1409 | 40006377 |