Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175060.2 | NP_778230.1 | 1 | 1417 | ||
GenBank mRNA | AF161403.1 | - | 1 | 371 | ||
AY358395.1 | - | 1 | 1382 | |||
AY573061.1 | - | 1 | 1096 | |||
AY606132.1 | - | 1 | 1142 | |||
BC031567.1 | - | 1 | 1096 | 5180301 | ||
BX248017.1 | - | 1 | 1416 |