Gene Symbol: PXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183373.3 | NP_899229.2 | 3 | 929 | ||
XM_011514393.2 | XP_011512695.1 | 5 | 908 | |||
GenBank mRNA | AK024828.1 | - | 3 | 402 | ||
AK289598.1 | - | 3 | 630 | |||
AL832792.1 | - | 3 | 475 | |||
BC045715.2 | - | 2 | 193 | |||
BC056908.1 | - | 3 | 619 | 6158428 |