Gene Symbol: PXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183373.3 | NP_899229.2 | 5 | 2084 | ||
XM_011514393.2 | XP_011512695.1 | 7 | 2063 | |||
GenBank mRNA | AJ420534.1 | - | 1 | 1489 | ||
AK024828.1 | - | 5 | 1557 | |||
AL832792.1 | - | 5 | 1629 | |||
BC045715.2 | - | 4 | 1348 | |||
BC056908.1 | - | 5 | 1773 | 6158428 |