Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198621.2 | NP_001185550.1 | 2 | 2702 | ||
NM_194293.2 | NP_919269.2 | 2 | 2692 | |||
XM_005264909.3 | XP_005264966.1 | 2 | 2651 | |||
GenBank mRNA | AJ626899.1 | - | 2 | 2700 | ||
AJ626900.1 | - | 2 | 2700 | |||
AK122723.1 | - | 1 | 99 | |||
AK126299.1 | - | 1 | 1142 | |||
AL713648.1 | - | 1 | 780 | |||
AL831956.1 | - | 1 | 487 | |||
AL832020.1 | - | 1 | 780 | |||
AL832459.1 | - | 1 | 780 | |||
AY375160.1 | - | 2 | 2692 | |||
CR749430.1 | - | 2 | 2702 |