Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001971.2 | NP_001001971.1 | 8 | 1026 | ||
NM_001143773.1 | NP_001137245.1 | 9 | 1197 | |||
NM_001166698.1 | NP_001160170.1 | 9 | 1111 | |||
NM_198215.3 | NP_937858.2 | 8 | 1026 | |||
XM_005269618.4 | XP_005269675.2 | 8 | 1026 | |||
XM_005269619.4 | XP_005269676.2 | 8 | 1026 | |||
XM_006717702.3 | XP_006717765.1 | 10 | 1027 | |||
XM_006717703.3 | XP_006717766.1 | 10 | 1209 | |||
XM_011539490.2 | XP_011537792.1 | 9 | 1082 | |||
XM_011539491.2 | XP_011537793.1 | 10 | 1128 | |||
XM_017015885.1 | XP_016871374.1 | 9 | 940 | |||
XM_017015886.1 | XP_016871375.1 | 9 | 1197 | |||
XM_017015887.1 | XP_016871376.1 | 9 | 947 | |||
XM_017015888.1 | XP_016871377.1 | 10 | 1209 | |||
XM_017015889.1 | XP_016871378.1 | 10 | 1027 | |||
XM_017015890.1 | XP_016871379.1 | 10 | 1266 | |||
XM_017015891.1 | XP_016871380.1 | 9 | 1197 | |||
XM_017015892.1 | XP_016871381.1 | 9 | 1197 | |||
XM_017015893.1 | XP_016871382.1 | 11 | 1298 | |||
GenBank mRNA | AK289796.1 | - | 8 | 952 | ||
AK294805.1 | - | 8 | 941 | |||
AK299924.1 | - | 9 | 1197 | |||
AK316542.1 | - | 9 | 1107 | |||
BC036453.1 | - | 8 | 984 | 5270033 | ||
BC064431.1 | - | 8 | 1026 | 5764753 | ||
U79304.1 | - | 6 | 634 |