Hamburger Menu Button
Thermo Fisher Scientific Logo
๋ก๊ทธ์ธ
ํ์์ด ์๋์ ๊ฐ์?
๊ณ์ ์์ฑํ๊ธฐ
๋ชจ๋ ์ ํ ์ฃผ๋ฌธ
ํญ์ฒด
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR ๋ถ์
ํผํซ ๋ฐ ํผํซ ํ
์คํ์ค์ฉ ์์ฌ๋ถ๋ฆฌ๊ธฐ
์ด์ ์จ ๋๋๊ณ
๋ถ๊ดํ
๋น์ด์ปค
PCR ์ฅ๋น ๋ฐ ์ฉํ
์ ํ ์นดํ ๊ณ ๋ฆฌ ์ ์ฒด๋ณด๊ธฐ
์์ฉ ๋ถ์ผ ๋ฐ ๊ธฐ๋ฒ
์ธํฌ ๋ถ์
์คํ์ค ์ฅ๋น
Real-Time PCR
PCR
ํฌ๋ก๋งํ ๊ทธ๋ํผ
์ธํฌ ๋ฐฐ์ ๋ฐ ํธ๋์คํ์
DNA/RNA ์ถ์ถ๊ณผ ๋ถ์
๋จ๋ฐฑ์ง ์๋ฌผํ
์ ์ธํฌ๋ถ์
ํํ ์ ํ
์์ฉ ๋ถ์ผ ๋ฐ ๊ธฐ๋ฒ ์ ์ฒด๋ณด๊ธฐ
์๋น์ค
๋ง์ถคํ ์๋น์ค
์ํฐํ๋ผ์ด์ฆ๊ธ ์คํ์ค ์ ๋ณด ์ฒ๋ฆฌ
์ํฐํ๋ผ์ด์ฆ ์๋น์ค
ํตํฉ CRO CDMO ์๋น์ค
CDMO ์๋น์ค
์์ ์ํ CRO ์๋น
์ฅ๋น ์๋น์ค
๊ต์ก ์๋น์ค
์๋น์ค ์ ์ฒด ๋ณด๊ธฐ
์ง์
๊ณ ๊ฐ์ผํฐ
๋ฌธ์ํ๊ธฐ
์ํ์ฑ์ ์(COA) ๋ฐ ์ ํฉ์ฑ ์ธ์ฆ์(COC)
Safety Data Sheets (SDS)
๋งค๋ด์ผ
์ธ์ฉ ๋ฐ ์ฐธ๊ณ ๋ฌธํ
๊ธฐ๊ธฐ ์ง์
๊ธฐ์ ์๋ฃ/์ ํ FAQ
ํ์ต ์ผํฐ
์ง์ ๋ฉ๋ด ์ ์ฒด๋ณด๊ธฐ
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
๋ฌธ์ํ๊ธฐ
๋น ๋ฅธ ์ฃผ๋ฌธ
์ฃผ๋ฌธ ํํฉ
์ ํ ๋ฌธ์ ๊ฒ์
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
์ ์ฒด๊ฒ์
์ ์ฒด๊ฒ์
Search button
Search button
Close
์ฃผ๋ฌธ ํํฉ
๋น ๋ฅธ์ฃผ๋ฌธ
๋ก๊ทธ์ธ
๋ก๊ทธ์ธ
ํ์์ด ์๋์ ๊ฐ์?
๊ณ์ ์์ฑํ๊ธฐ
๊ณ์ ์ ๋ณด
์ฃผ๋ฌธ๋ด์ญ์กฐํ
์ปค์คํ ์ ํ ๋ฐ ํ๋ก์ ํธ
์ฅ๋น๊ด๋ฆฌ
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNK1 SIRNA Assays ›
Gene Symbol:
WNK1
Gene Name:
WNK lysine deficient protein kinase 1
Gene Aliases:
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
Chromosome Location:
Chr.12: 752552 - 911452 on Build GRCh38
UniGene ID:
Hs.744906
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130233
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WNK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184985.1
NP_001171914.1
2
1456
NM_014823.2
NP_055638.2
2
1456
NM_018979.3
NP_061852.3
2
1456
NM_213655.4
NP_998820.3
2
1456
XM_006719003.2
XP_006719066.1
2
1781
XM_011520997.2
XP_011519299.1
2
1827
XM_011520998.2
XP_011519300.1
2
1781
XM_011520999.2
XP_011519301.1
2
1781
XM_011521000.2
XP_011519302.1
2
1781
XM_011521001.2
XP_011519303.1
2
1781
XM_011521002.2
XP_011519304.1
2
1781
XM_011521003.2
XP_011519305.1
2
1781
XM_011521004.2
XP_011519306.1
2
1781
XM_011521006.2
XP_011519308.1
2
1781
XM_011521007.2
XP_011519309.1
2
1781
XM_011521008.2
XP_011519310.1
2
1781
XM_011521009.2
XP_011519311.1
2
1781
XM_017019834.1
XP_016875323.1
2
1781
XM_017019835.1
XP_016875324.1
2
1781
XM_017019836.1
XP_016875325.1
2
1781
XM_017019837.1
XP_016875326.1
2
1781
XM_017019838.1
XP_016875327.1
2
1781
XM_017019839.1
XP_016875328.1
2
1781
GenBank mRNA
AB002342.2
-
2
611
AF061944.1
-
2
1456
AJ296290.1
-
2
813
JQ358908.1
-
2
813
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of systemic arterial blood pressure
protein phosphorylation
ion transport
negative regulation of phosphatase activity
peptidyl-threonine phosphorylation
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
protein autophosphorylation
neuron development
regulation of cellular process
negative regulation of protein serine/threonine kinase activity
negative regulation of pancreatic juice secretion
regulation of ion homeostasis
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
chloride channel inhibitor activity
potassium channel inhibitor activity
protein kinase binding
phosphatase binding
protein serine/threonine kinase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0