Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020461.3 | NP_065194.2 | 7 | 2065 | ||
GenBank mRNA | AB051456.1 | - | 7 | 1459 | ||
AF272887.1 | - | 7 | 1658 | |||
AL832617.2 | - | 7 | 2086 | |||
BC144488.1 | - | 7 | 1657 | 9053015 | ||
BC144489.1 | - | 7 | 1657 | |||
BC150613.1 | - | 7 | 1657 | 9056983 | ||
BC151209.1 | - | 7 | 2047 | 40147276 |