Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205319.1 | NP_001192248.1 | 8 | 1566 | ||
NM_022463.4 | NP_071908.2 | 8 | 1909 | |||
XM_005256756.4 | XP_005256813.1 | 7 | 1735 | |||
XM_005256758.3 | XP_005256815.1 | 8 | 2238 | |||
GenBank mRNA | AK022676.1 | - | 6 | 1338 | ||
AK027451.1 | - | 7 | 1462 | |||
AY189690.1 | - | 7 | 1449 | |||
BC063828.1 | - | 5 | 1164 | |||
BC073845.1 | - | 6 | 1319 |