Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022475.2 | NP_071920.1 | 10 | 2321 | ||
XM_005263178.4 | XP_005263235.1 | 10 | 2190 | |||
XM_006714288.3 | XP_006714351.1 | 10 | 2190 | |||
GenBank mRNA | AK024645.1 | - | 3 | 308 | ||
AK291849.1 | - | 10 | 1661 | |||
AY009317.1 | - | 10 | 2161 | |||
AY009951.1 | - | 10 | 2296 | |||
AY358747.1 | - | 10 | 2271 | |||
BC025311.1 | - | 10 | 2147 | 4122618 |