Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022749.5 | NP_073586.5 | 17 | 3711 | ||
XM_006716381.2 | XP_006716444.1 | 17 | 3741 | |||
XM_006716385.3 | XP_006716448.1 | 18 | 5236 | |||
XM_017013732.1 | XP_016869221.1 | 17 | 3708 | |||
XM_017013733.1 | XP_016869222.1 | 18 | 3828 | |||
XM_017013734.1 | XP_016869223.1 | 17 | 3692 | |||
GenBank mRNA | AF318332.1 | - | 1 | 1403 | ||
AF495722.1 | - | 12 | 3117 | |||
AK025411.1 | - | 3 | 1685 | |||
AK025454.1 | - | 6 | 2121 | |||
AK055191.1 | - | 11 | 2770 | |||
AK095924.1 | - | 4 | 2324 | |||
BC012865.1 | - | 8 | 2524 | |||
BC013350.2 | - | 7 | 2202 | |||
BC052237.1 | - | 18 | 2825 |