Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM184A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100411.2 | NP_001093881.1 | 13 | 2554 | ||
NM_001288576.1 | NP_001275505.1 | 13 | 2554 | |||
NM_024581.5 | NP_078857.5 | 13 | 2945 | |||
GenBank mRNA | AK024004.1 | - | 6 | 749 | ||
AK091682.1 | - | 13 | 2550 | |||
AK127744.1 | - | 10 | 1394 | |||
AK308316.1 | - | 13 | 3041 | |||
AL136767.1 | - | 2 | 60 | |||
AY131226.1 | - | 9 | 1221 | |||
BC009055.1 | - | 5 | 537 | |||
BC060769.1 | - | 13 | 2945 | 30337859 | ||
BX538186.1 | - | 13 | 2534 |