Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024644.4 | NP_078920.2 | 1 | 1149 | ||
GenBank mRNA | AB101204.1 | - | 1 | 1256 | ||
AK025455.1 | - | 1 | 1119 | |||
AK299994.1 | - | 2 | 810 | |||
AY390535.1 | - | 1 | 1116 | |||
BC011350.1 | - | 1 | 1118 | 4340013 | ||
BC071954.1 | - | 1 | 1096 | 6728391 | ||
BX390965.1 | - | 1 | 300 |