Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320210.1 | NP_001307139.1 | 7 | 839 | ||
NM_001321238.1 | NP_001308167.1 | 5 | 703 | |||
NM_001321239.1 | NP_001308168.1 | 5 | 1553 | |||
NM_024818.4 | NP_079094.1 | 7 | 1689 | |||
NM_198329.3 | NP_938143.1 | 7 | 712 | |||
GenBank mRNA | AB154406.1 | - | 7 | 648 | ||
AK026904.1 | - | 7 | 882 | |||
AK027563.1 | - | 6 | 481 | |||
AK056238.1 | - | 7 | 1690 | |||
AK129616.1 | - | 8 | 748 | |||
AL136757.1 | - | 7 | 702 | |||
AL832669.1 | - | 4 | 1908 | |||
AY253672.1 | - | 7 | 841 | |||
BC009737.1 | - | 7 | 818 | 3879061 | ||
BC048015.1 | - | 7 | 796 |