Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320210.1 | NP_001307139.1 | 12 | 1827 | ||
NM_001321238.1 | NP_001308167.1 | 10 | 1691 | |||
NM_001321239.1 | NP_001308168.1 | 10 | 2541 | |||
NM_024818.4 | NP_079094.1 | 12 | 2677 | |||
NM_198329.3 | NP_938143.1 | 12 | 1700 | |||
GenBank mRNA | AK026904.1 | - | 12 | 1870 | ||
AK027563.1 | - | 11 | 1469 | |||
AL136757.1 | - | 12 | 1690 | |||
AL832669.1 | - | 8 | 3063 | |||
AY253672.1 | - | 12 | 1829 | |||
BC048015.1 | - | 12 | 1784 |