Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320210.1 | NP_001307139.1 | 12 | 2271 | ||
NM_001321238.1 | NP_001308167.1 | 10 | 2135 | |||
NM_001321239.1 | NP_001308168.1 | 10 | 2985 | |||
NM_024818.4 | NP_079094.1 | 12 | 3121 | |||
NM_198329.3 | NP_938143.1 | 12 | 2144 | |||
GenBank mRNA | AL832669.1 | - | 8 | 3507 | ||
AY253672.1 | - | 12 | 2273 | |||
BC048015.1 | - | 12 | 2228 |