Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TXNRD1 SIRNA Assays ›
Gene Symbol:
TXNRD1
Gene Name:
thioredoxin reductase 1
Gene Aliases:
GRIM-12, TR, TR1, TRXR1, TXNR
Chromosome Location:
Chr.12: 104215779 - 104350307 on Build GRCh38
UniGene ID:
Hs.654922
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13077
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TXNRD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001093771.2
NP_001087240.1
7
651
NM_001261445.1
NP_001248374.1
5
1073
NM_001261446.1
NP_001248375.1
4
628
NM_003330.3
NP_003321.3
5
988
NM_182729.2
NP_877393.1
5
751
NM_182742.2
NP_877419.1
4
878
NM_182743.2
NP_877420.1
4
641
GenBank mRNA
AF077367.1
-
4
305
AF208018.1
-
3
201
AJ001050.1
-
3
214
AK128515.1
-
4
536
AK226071.1
-
5
459
AK293322.1
-
4
288
AK295067.1
-
5
387
AK296495.1
-
4
278
AK304241.1
-
7
634
AK308881.1
-
5
682
AK312262.1
-
5
650
AY057105.1
-
7
627
AY344080.1
-
5
372
AY344081.1
-
5
421
AY344083.1
-
5
538
AY344086.1
-
5
377
AY344087.1
-
5
442
AY344089.1
-
5
393
AY344092.1
-
5
380
AY344093.1
-
5
364
AY344096.1
-
4
318
AY344669.1
-
4
249
AY344670.1
-
4
373
AY344672.1
-
4
338
AY344673.1
-
4
267
AY344675.1
-
4
236
AY344679.1
-
4
301
BC018122.1
-
5
364
3883490
BT019640.1
-
3
177
CR536506.1
-
3
177
D88687.1
-
5
429
DA319870.1
-
4
428
DA440625.1
-
5
453
S79851.1
-
5
616
X91247.1
-
5
616
Back To Top
More Information
Panther Classification:
Molecular Function -
reductase
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
mesoderm formation
selenium compound metabolic process
signal transduction
cell proliferation
nucleobase-containing small molecule interconversion
cell redox homeostasis
oxidation-reduction process
cellular oxidant detoxification
thioredoxin-disulfide reductase activity
protein binding
electron carrier activity
protein disulfide oxidoreductase activity
flavin adenine dinucleotide binding
methylselenol reductase activity
methylseleninic acid reductase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0