Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270367.1 | NP_001257296.1 | 4 | 762 | ||
NM_001270368.1 | NP_001257297.1 | 4 | 775 | |||
NM_001270369.1 | NP_001257298.1 | 5 | 814 | |||
NM_001270370.1 | NP_001257299.1 | 6 | 821 | |||
NM_001270371.1 | NP_001257300.1 | 5 | 836 | |||
NM_001270372.1 | NP_001257301.1 | 4 | 754 | |||
NM_001270373.1 | NP_001257302.1 | 5 | 793 | |||
NM_001270374.1 | NP_001257303.1 | 5 | 1143 | |||
NM_025125.3 | NP_079401.2 | 4 | 741 | |||
NR_072984.1 | 5 | 758 | ||||
NR_072985.1 | 5 | 765 | ||||
NR_072986.1 | 4 | 663 | ||||
NR_072987.1 | 5 | 715 | ||||
XM_006717978.2 | XP_006718041.1 | 5 | 1093 | |||
GenBank mRNA | AK023325.1 | - | 4 | 717 | ||
AK222536.1 | - | 4 | 722 | |||
AK300490.1 | - | 4 | 772 | |||
AK309980.1 | - | 4 | 749 | |||
AK310160.1 | - | 5 | 723 | |||
AK311563.1 | - | 5 | 731 | |||
AL832397.1 | - | 4 | 638 | |||
AL832483.1 | - | 4 | 749 | |||
BC022252.1 | - | 4 | 717 | 4668550 | ||
BX537803.1 | - | 5 | 723 |