Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201573.1 | NP_001188502.1 | 8 | 658 | ||
NM_001201574.1 | NP_001188503.1 | 5 | 561 | |||
NM_025152.2 | NP_079428.2 | 10 | 894 | |||
NR_120408.1 | 9 | 800 | ||||
XM_011537181.2 | XP_011535483.1 | 7 | 829 | |||
XM_011537182.2 | XP_011535484.1 | 11 | 868 | |||
XM_011537184.2 | XP_011535486.1 | 5 | 744 | |||
XM_017021667.1 | XP_016877156.1 | 9 | 815 | |||
GenBank mRNA | AK022722.1 | - | 10 | 807 | ||
AK093789.1 | - | 5 | 561 | |||
AK295326.1 | - | 10 | 855 | |||
AK301447.1 | - | 8 | 658 | |||
AK316445.1 | - | 10 | 884 | |||
BC024919.1 | - | 10 | 820 | 4475196 | ||
BP274976.1 | - | 5 | 492 | |||
BX248028.1 | - | 9 | 787 | |||
BX648419.1 | - | 2 | 2874 | |||
KJ534909.1 | - | 10 | 839 |