Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC44A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178044.1 | NP_001171515.1 | 7, 8 | 565 | ||
NM_001178045.1 | NP_001171516.1 | 8, 9 | 694 | |||
NM_025257.2 | NP_079533.2 | 8, 9 | 691 | |||
GenBank mRNA | AK027397.1 | - | 8, 9 | 642 | ||
AK222998.1 | - | 8, 9 | 642 | |||
AK223013.1 | - | 8, 9 | 642 | |||
AK300550.1 | - | 8, 9 | 694 | |||
AK301596.1 | - | 7, 8 | 516 | |||
AK314673.1 | - | 8, 9 | 646 | |||
AL833009.1 | - | 4, 5 | 384 | |||
BC014659.1 | - | 8, 9 | 630 | 4644272 | ||
DA924720.1 | - | 1, 2 | 95 | |||
KU175229.1 | - | 8, 9 | 625 |