Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178044.1 | NP_001171515.1 | 20 | 2136 | ||
NM_001178045.1 | NP_001171516.1 | 21 | 2265 | |||
NM_025257.2 | NP_079533.2 | 21 | 2262 | |||
GenBank mRNA | AK027397.1 | - | 21 | 2213 | ||
AK222998.1 | - | 21 | 2213 | |||
AK223013.1 | - | 12 | 1127 | |||
AK300550.1 | - | 21 | 2265 | |||
AK301596.1 | - | 20 | 2087 | |||
AL833009.1 | - | 17 | 1955 | |||
AY358457.1 | - | 9 | 1029 | |||
BC014659.1 | - | 21 | 2198 | 4644272 |