Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135947.2 | NP_001129419.1 | 4 | 772 | ||
NM_001265582.1 | NP_001252511.1 | 3 | 2135 | |||
NM_030914.3 | NP_112176.1 | 5 | 416 | |||
NR_049743.1 | 5 | 457 | ||||
GenBank mRNA | AK001880.1 | - | 5 | 365 | ||
AK097029.1 | - | 3 | 2081 | |||
AK300877.1 | - | 4 | 719 | |||
BC003581.1 | - | 5 | 374 | 3546655 | ||
BI823604.1 | - | 5 | 415 | |||
D83200.1 | - | 4 | 321 | |||
DA691216.1 | - | 5 | 422 |