Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030918.5 | NP_112180.4 | 2 | 441 | ||
XM_005245509.2 | XP_005245566.1 | 2 | 563 | |||
XM_005245510.3 | XP_005245567.1 | 2 | 173 | |||
XM_005245511.4 | XP_005245568.1 | 2 | 469 | |||
XM_011510026.2 | XP_011508328.1 | 2 | 537 | |||
GenBank mRNA | AB007957.2 | - | 2 | 410 | ||
AY044866.1 | - | 1 | 146 | |||
BC012184.1 | - | 2 | 431 | |||
BC100998.1 | - | 2 | 441 | 40006277 | ||
BC100999.1 | - | 2 | 439 | 40006279 | ||
BC101000.1 | - | 2 | 441 | 40006281 | ||
BC101822.1 | - | 2 | 476 | 8069328 | ||
BC101824.1 | - | 2 | 476 | 8069330 | ||
BG472531.1 | - | 2 | 413 | |||
BM560557.1 | - | 441 |