Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030919.2 | NP_112181.3 | 3 | 839 | ||
XM_017028088.1 | XP_016883577.1 | 3 | 722 | |||
GenBank mRNA | AK055793.1 | - | 3 | 841 | ||
AK303860.1 | - | 4 | 686 | |||
BC001068.2 | - | 3 | 725 | |||
BC006553.2 | - | 3 | 725 | |||
BC053683.1 | - | 3 | 726 | |||
BC063661.1 | - | 3 | 725 | |||
BC080188.1 | - | 3 | 726 |