Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030940.3 | NP_112202.2 | 4 | 1869 | ||
GenBank mRNA | AF038186.1 | - | 1 | 1284 | ||
AF284752.1 | - | 4 | 1834 | |||
AK055643.1 | - | 4 | 1832 | |||
AK225937.1 | - | 4 | 1832 | |||
AL834356.1 | - | 4 | 1814 | |||
BC002675.2 | - | 4 | 1822 | 3608301 | ||
BC053900.1 | - | 4 | 1790 | |||
BC071621.1 | - | 4 | 1803 | 5107151 |