Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC00597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026813.1 | 1 | 460 | |||
GenBank mRNA | AF216224.1 | - | 1 | 466 | ||
AL109679.1 | - | 1 | 105 | |||
BC069765.1 | - | 1 | 396 | |||
BC103988.1 | - | 1 | 386 | |||
BC103989.1 | - | 1 | 386 | |||
BC103990.1 | - | 1 | 386 | |||
BC103991.1 | - | 1 | 386 |