Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF146 SIRNA Assays ›
Gene Symbol:
RNF146
Gene Name:
ring finger protein 146
Gene Aliases:
-
Chromosome Location:
Chr.6: 127266340 - 127289893 on Build GRCh38
UniGene ID:
Hs.267120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131065
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF146
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242844.1
NP_001229773.1
5
809
NM_001242845.1
NP_001229774.1
5
658
NM_001242846.1
NP_001229775.1
5
571
NM_001242847.1
NP_001229776.1
4
700
NM_001242848.1
NP_001229777.1
4
430
NM_001242849.1
NP_001229778.1
3
572
NM_001242850.1
NP_001229779.1
3
302
NM_001242851.1
NP_001229780.1
3
242
NM_001242852.1
NP_001229781.1
5
789
NM_030963.3
NP_112225.2
5
539
XM_006715571.3
XP_006715634.1
5
1239
XM_011536161.2
XP_011534463.1
6
1126
XM_011536162.2
XP_011534464.1
6
1178
XM_011536163.2
XP_011534465.1
6
1158
XM_011536164.2
XP_011534466.1
5
1017
XM_017011336.1
XP_016866825.1
3
919
XM_017011337.1
XP_016866826.1
5
4160
XM_017011338.1
XP_016866827.1
7
1267
XM_017011339.1
XP_016866828.1
6
1296
XM_017011340.1
XP_016866829.1
6
1348
XM_017011341.1
XP_016866830.1
4
1047
XM_017011342.1
XP_016866831.1
5
1188
XM_017011343.1
XP_016866832.1
6
1297
GenBank mRNA
AJ315122.1
-
1
104
AJ420549.1
-
2
174
AK027436.1
-
2
243
AK027558.1
-
3
291
AK027776.1
-
1
114
AL136829.1
-
5
502
BC008235.1
-
2
191
3868592
BG708628.1
-
3
537
BI771613.1
-
2
241
CN388414.1
-
3
382
CR533514.1
-
1
103
DA043259.1
-
5
527
DA145911.1
-
4
417
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Wnt signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein autoubiquitination
protein K48-linked ubiquitination
positive regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
poly-ADP-D-ribose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0