Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297602.1 | NP_001284531.1 | 4, 5 | 1379 | ||
NM_031299.5 | NP_112589.1 | 4, 5 | 1454 | |||
GenBank mRNA | AK291421.1 | - | 4, 5 | 657 | ||
AK312843.1 | - | 4, 5 | 674 | |||
BC002551.2 | - | 4, 5 | 649 | 3138951 | ||
BM911472.1 | - | 4, 5 | 564 |