Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017440.2 | NP_001017440.1 | 6 | 1970 | ||
NM_031468.3 | NP_113656.2 | 7 | 1960 | |||
XM_011516594.2 | XP_011514896.1 | 6 | 1812 | |||
XM_011516595.1 | XP_011514897.1 | 6 | 1560 | |||
XM_011516596.2 | XP_011514898.1 | 7 | 2455 | |||
XM_011516597.2 | XP_011514899.1 | 6 | 2388 | |||
XM_011516599.1 | XP_011514901.1 | 6 | 1759 | |||
XM_017012675.1 | XP_016868164.1 | 8 | 2315 | |||
XM_017012676.1 | XP_016868165.1 | 7 | 2097 | |||
XM_017012677.1 | XP_016868166.1 | 6 | 1610 | |||
XM_017012678.1 | XP_016868167.1 | 7 | 2072 | |||
XM_017012679.1 | XP_016868168.1 | 6 | 2425 | |||
XM_017012680.1 | XP_016868169.1 | 6 | 1724 | |||
XM_017012681.1 | XP_016868170.1 | 6 | 1587 | |||
XM_017012682.1 | XP_016868171.1 | 6 | 2137 | |||
XM_017012683.1 | XP_016868172.1 | 7 | 1921 | |||
GenBank mRNA | AF282250.1 | - | 7 | 1692 | ||
AK091485.1 | - | 1 | 485 | |||
AK123862.1 | - | 6 | 1506 |