Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GORASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278789.1 | NP_001265718.1 | 7 | 1637 | ||
NM_001278790.1 | NP_001265719.1 | 6 | 1457 | |||
NM_031899.3 | NP_114105.1 | 9 | 2385 | |||
NR_103866.1 | 9 | 1881 | ||||
NR_103867.1 | 9 | 1873 | ||||
XM_006713301.3 | XP_006713364.1 | 9 | 2485 | |||
XM_011534020.2 | XP_011532322.1 | 9 | 2395 | |||
XM_011534021.2 | XP_011532323.1 | 8 | 2305 | |||
XM_017007050.1 | XP_016862539.1 | 9 | 1571 | |||
XM_017007051.1 | XP_016862540.1 | 8 | 1920 | |||
XM_017007052.1 | XP_016862541.1 | 8 | 1516 | |||
GenBank mRNA | AJ409349.1 | - | 9 | 1661 | ||
AK001574.1 | - | 8 | 1903 | |||
AK027096.1 | - | 53 | ||||
AK057043.1 | - | 6 | 1195 | |||
AK091168.1 | - | 9 | 1809 | |||
AK124755.1 | - | 9 | 2385 | |||
BC008928.2 | - | 9 | 1602 | |||
BC075854.1 | - | 9 | 1671 | 5503263 | ||
BC103826.1 | - | 9 | 1603 | 40001584 | ||
BC103827.1 | - | 9 | 1603 | 40001588 |